Cadmium ≥99.99% (metals basis), powder -200 mesh
Supplier: Thermo Fisher Scientific
|
Danger
|
11353.06
11353.14
11353.22
11353.06EA
102
EUR
11353.06
11353.14
11353.22
Cadmium ≥99.99% (metals basis), powder -200 mesh
Cadmium
Purity: 99.99%
CAS No.: 7440-43-9
Appearance: Silvery white
UN No.: 3179
MDL No.: MFCD00010914
Formula:
Cd Boiling Pt: 767 °C (1013 hPa) Melting Pt: 321 °C Density: 8.64 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010914 CAS Number: 7440-43-9 EINECS: 231-152-8 UN: 3179 ADR: 4.1,II |