Cadmium ≥99.5% (metals basis), powder -325 mesh
Supplier: Thermo Fisher Scientific
Danger
11891.09 11891.22 11891.36
11891.09EA 40.9 EUR
11891.09 11891.22 11891.36
Cadmium ≥99.5% (metals basis), powder -325 mesh
Cadmium
Purity: 99.5% CAS No.: 7440-43-9 Appearance: Silver white UN No.: 3179 MDL No.: MFCD00010914
Formula: Cd
Boiling Pt: 767 °C (1013 hPa)
Melting Pt: 321 °C
Density: 8.64 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010914
CAS Number: 7440-43-9
EINECS: 231-152-8
UN: 3179
ADR: 4.1,II