Mercury
Supplier: Roth Carl
|
Danger
|
7594.1
7594.2
ROTH7594.1EA
267
EUR
ROTH7594.1
ROTH7594.2
Mercury
Mercury
Formula:
Hg Boiling Pt: 356.5 °C (1013 hPa) Melting Pt: –39 °C Density: 13.54 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011035 CAS Number: 7439-97-6 EINECS: 231-106-7 UN: 2809 ADR: 8,III Merck Index: 13,05925 |