Zinc, powder, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
324930-10G 324930-50G
SIAL324930-10GEA 289 EUR
SIAL324930-10G SIAL324930-50G
Zinc, powder, Sigma-Aldrich®
Zinc
Formula: Zn
Boiling Pt: 907 °C (1013 hPa)
Melting Pt: 419.5 °C
Density: 7.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
UN: 1436
ADR: 4.3,II