Hafnium ≥99.97% (metals basis excluding Zr), wire (Zr ∼ 3%), Ø 0.25 mm (0.01 in)
Supplier: Thermo Scientific
10200.G2 10200.G6
10200.G2EA 207 EUR
10200.G2 10200.G6
Hafnium ≥99.97% (metals basis excluding Zr), wire (Zr ∼ 3%), Ø 0.25 mm (0.01 in)
Hafnium
Purity: 99.97% CAS No.: 7440-58-6 Physical Form: ≈0.64g/m Odour: Odourless MDL No.: MFCD00011032
Formula: Hf
MW: 178.49 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011032
CAS Number: 7440-58-6
EINECS: 231-166-4