Hafnium ≥99.95% (metals basis excluding Zr), wire (Zr ∼ 3%), Ø 1.0 mm (0.04 in)
Supplier: Thermo Scientific
10205.BU 10205.BY 10205.CC
10205.BUEA 86.6 EUR
10205.BU 10205.BY 10205.CC
Hafnium ≥99.95% (metals basis excluding Zr), wire (Zr ∼ 3%), Ø 1.0 mm (0.04 in)
Hafnium
Purity: 99.95% CAS No.: 7440-58-6 Physical Form: ≈10.28g/m Odour: Odourless MDL No.: MFCD00011032
Formula: Hf
MW: 178.49 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011032
CAS Number: 7440-58-6
EINECS: 231-166-4