Titanium ≥99.98% (metals basis), wire, Ø 0.5 mm (0.02 in)
Supplier: Thermo Fisher Scientific
Warning
00362.G1 00362.G5
00362.G1EA 198 EUR
00362.G1 00362.G5
Titanium ≥99.98% (metals basis), wire, Ø 0.5 mm (0.02 in)
Titanium
Purity: 99.98% CAS No.: 7440-32-6 Physical Form: ≈0.88g/m Odour: Odourless MDL No.: MFCD00011264
Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3262
Melting Pt: 1660
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610