Titanium 99,5 Metals basis, foil, annealed, Thickness: 0.25 mm (0.01 in)
Supplier: Thermo Fisher Scientific
Warning
10385.GH 10385.HP 10385.HY
10385.GHEA 92.4 EUR
10385.GH 10385.HP 10385.HY
Titanium 99,5 Metals basis, foil, annealed, Thickness: 0.25 mm (0.01 in)
Titanium
Purity: 99.5% CAS No.: 7440-32-6 Physical Form: ≈11.3g/100x100mm Odour: Odourless MDL No.: MFCD00011264
Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3262
Melting Pt: 1660
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610