Titanium ≥99.7% (metals basis), rods, Ø 6.4 mm (0.25 in)
Supplier: Thermo Fisher Scientific
Warning
10393.BU 10393.BY 10393.CQ
10393.BUEA 49.7 EUR
10393.BU 10393.BY 10393.CQ
Titanium ≥99.7% (metals basis), rods, Ø 6.4 mm (0.25 in)
Titanium
Purity: 99.7% CAS No.: 7440-32-6 Physical Form: ≈1.45g/cm Odour: Odourless MDL No.: MFCD00011264
Formula: Ti
Boiling Pt: 3262 °C (1013 hPa)
Melting Pt: 1660 °C
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610