Titanium 99,7 Metals basis, wire, annealed, Length: 90 cm (35 in), Ø 0.81 mm (0.032 in)
Supplier: Thermo Fisher Scientific
|
Warning
|
10395.KD
10395.KK
10395.KDEA
647
EUR
10395.KD
10395.KK
Titanium 99,7 Metals basis, wire, annealed, Length: 90 cm (35 in), Ø 0.81 mm (0.032 in)
Titanium
Purity: 99.7%
CAS No.: 7440-32-6
Physical Form: ≈2.32g/m, Temper: annealed
Odour: Odourless
MDL No.: MFCD00011264
Formula:
Ti MW: 47.88 g/mol Boiling Pt: 3262 Melting Pt: 1660 Density: 4.51 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011264 CAS Number: 7440-32-6 EINECS: 231-142-3 Merck Index: 12,09610 |