Titanium ≥99.7% (metals basis), wire, Ø 0.25 mm (0.01 in)
Supplier: Thermo Fisher Scientific
Warning
10396.G6 10396.H2
10396.G6EA 60.6 EUR
10396.G6 10396.H2
Titanium ≥99.7% (metals basis), wire, Ø 0.25 mm (0.01 in)
Titanium
Purity: 99.7% CAS No.: 7440-32-6 Physical Form: ≈0.22g/m, Temper: as drawn Odour: Odourless MDL No.: MFCD00011264
Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3262
Melting Pt: 1660
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610