Titanium ≥99.99% (metals basis), foil, Thickness: 0.127 mm (0.005 in)
Supplier: Thermo Scientific
Warning
13976.FF 13976.FI 13976.GH
13976.FFEA 73.9 EUR
13976.FF 13976.FI 13976.GH
Titanium ≥99.99% (metals basis), foil, Thickness: 0.127 mm (0.005 in)
Titanium
Purity: 99.99+% CAS No.: 7440-32-6 Physical Form: ≈0.36g/25x25mm Odour: Odourless MDL No.: MFCD00011264
Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3262 °C (1013 hPa)
Melting Pt: 1660 °C
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610