Titanium ≥99.99% (metals basis), rods, Ø 6.4 mm (0.25 in)
Supplier: Thermo Fisher Scientific
Warning
14001.BQ 14001.BU 14001.CZ
14001.BQEA 105 EUR
14001.BQ 14001.BU 14001.CZ
Titanium ≥99.99% (metals basis), rods, Ø 6.4 mm (0.25 in)
Titanium
Purity: 99.99% CAS No.: 7440-32-6 Physical Form: ≈1.45g/cm Odour: Odourless MDL No.: MFCD00011264
Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3262
Melting Pt: 1660
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610