Titanium, Sigma-Aldrich®
Supplier: Merck
Warning
MQ100
369489-200G 369489-90G
SIAL369489-200GEA 212 EUR
SIAL369489-200G SIAL369489-90G
Titanium, Sigma-Aldrich®
Titanium
Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3262
Melting Pt: 1660
Density: 4.51 g/cm³ (20 °C)
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610