Titanium 99,4, powder -100 mesh
Supplier: Thermo Fisher Scientific
Warning
10383.14 10383.22 10383.36
10383.14EA 54.9 EUR
10383.14 10383.22 10383.36
Titanium 99,4, powder -100 mesh
Titanium
Purity: 99.4% CAS No.: 7440-32-6 Appearance: Grey Odour: Odourless UN No.: 2546 MDL No.: MFCD00011264
Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3262
Melting Pt: 1660
Density: 4.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
UN: 2546
ADR: 4.2 III
Merck Index: 12,09610