Titanium ≥99.5% (metals basis), powder -325 mesh
Supplier: Thermo Fisher Scientific
Danger
42624.22 42624.36 42624.D3
42624.22EA 140 EUR
42624.22 42624.36 42624.D3
Titanium ≥99.5% (metals basis), powder -325 mesh
Titanium
Purity: 99.5% CAS No.: 7440-32-6 Odour: Odourless UN No.: 2546 MDL No.: MFCD00011264
Formula: Ti
Boiling Pt: 3277 °C (1013 hPa)
Melting Pt: 1677 °C
Density: 4.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
UN: 2546
ADR: 4.2,II