Titanium, powder, Sigma-Aldrich®
Supplier: Merck
Danger
MQ100
366994-10G 366994-50G
SIAL366994-10GEA 256 EUR
SIAL366994-10G SIAL366994-50G
Titanium, powder, Sigma-Aldrich®
Titanium
Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3262
Melting Pt: 1660
Density: 4.5 g/cm³ (20 °C)
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
UN: 2546
ADR: 4.2 II
Merck Index: 12,09610