Niobium ≥99.8% (metals basis), foil, annealed, Thickness: 0.5 mm (0.02 in)
Supplier: Thermo Fisher Scientific
14358.FY 14358.HP 14358.HL
14358.FYEA 137 EUR
14358.FY 14358.HP 14358.HL
Niobium ≥99.8% (metals basis), foil, annealed, Thickness: 0.5 mm (0.02 in)
Niobium
Purity: 99.8% CAS No.: 7440-03-1 Physical Form: ≈21.4g/50x100mm Odour: Odourless MDL No.: MFCD00011126
Formula: Nb
MW: 92.91 g/mol
Boiling Pt: 3700
Melting Pt: 2468
Density: 8.57 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011126
CAS Number: 7440-03-1
EINECS: 231-113-5