Niobium ≥99.95% (metals basis excluding Ta), slug, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Supplier: Thermo Fisher Scientific
42847.14 42847.22
42847.14EA 102 EUR
42847.14 42847.22
Niobium ≥99.95% (metals basis excluding Ta), slug, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Niobium
Purity: 99.95% CAS No.: 7440-03-1 Physical Form: ≈1.7g/slug Odour: Odourless MDL No.: MFCD00011126
Formula: Nb
MW: 92.91 g/mol
Boiling Pt: 3700
Melting Pt: 2468
Density: 8.57 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011126
CAS Number: 7440-03-1
EINECS: 231-113-5