Niobium ≥99.8% (metals basis), powder -325 mesh
Supplier: Thermo Fisher Scientific
Danger
10275.14 10275.22 10275.36 10275.88
10275.14EA 95.7 EUR
10275.14 10275.22 10275.36 10275.88
Niobium ≥99.8% (metals basis), powder -325 mesh
Niobium
Purity: 99.8% CAS No.: 7440-03-1 Odour: Odourless UN No.: 3089 MDL No.: MFCD00011126
Formula: Nb
MW: 92.91 g/mol
Boiling Pt: 3700
Melting Pt: 2468
Storage Temperature: Ambient
MDL Number: MFCD00011126
CAS Number: 7440-03-1
EINECS: 231-113-5
UN: 3089
ADR: 4.1 II