Vanadium ≥99.8% (metals basis), slug, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Supplier: Thermo Fisher Scientific
42830.09 42830.18
42830.09EA 203 EUR
42830.09 42830.18
Vanadium ≥99.8% (metals basis), slug, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Vanadium
Purity: 99.8% CAS No.: 7440-62-2 Physical Form: ≈1.2g/slug Odour: Odourless MDL No.: MFCD00011453
Formula: V
Boiling Pt: 3477 °C (1013 hPa)
Melting Pt: 1896 °C
Density: 6.015 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011453
CAS Number: 7440-62-2
EINECS: 231-171-1
Merck Index: 12,10053