Vanadium, powder, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

262935-10G 262935-50G
SIAL262935-10GEA 194 EUR
SIAL262935-10G SIAL262935-50G
Vanadium, powder, Sigma-Aldrich®
Vanadium
Formula: V
Density: 6.11 g/cm³ (187 °C)
MDL Number: MFCD00011453
CAS Number: 7440-62-2
EINECS: 231-171-1
UN: 3089
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR