Molybdenum ≥99.95% (metals basis), powder -100 mesh
Supplier: Thermo Fisher Scientific
Warning
10031.18 10031.30 10031.A1
10031.18EA 59.4 EUR
10031.18 10031.30 10031.A1
Molybdenum ≥99.95% (metals basis), powder -100 mesh
Molybdenum
Purity: 99.95% CAS No.: 7439-98-7 Odour: Odourless UN No.: 3089 MDL No.: MFCD00003465
Formula: Mo
MW: 95.94 g/mol
Boiling Pt: 3617
Melting Pt: 2622
Density: 10.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003465
CAS Number: 7439-98-7
EINECS: 231-107-2
UN: 3089
ADR: 4.1 III
Merck Index: 12,06317