Molybdenum ≥99.95% (metals basis), wire, annealed, Ø 0.25 mm (0.01 in)
Supplier: Thermo Scientific
10036.G9 10036.H4
10036.G9EA 72.1 EUR
10036.G9 10036.H4
Molybdenum ≥99.95% (metals basis), wire, annealed, Ø 0.25 mm (0.01 in)
Molybdenum
Purity: 99.95% CAS No.: 7439-98-7 Physical Form: ≈0.50g/m, Temper: soft MDL No.: MFCD00003465
Formula: Mo
MW: 95.94 g/mol
Boiling Pt: 3617 °C (1013 hPa)
Melting Pt: 2622 °C
Density: 10.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003465
CAS Number: 7439-98-7
EINECS: 231-107-2
Merck Index: 12,06317