Molybdenum ≥99.95% (metals basis), foil, Thickness: 0.127 mm (0.005 in)
Supplier: Thermo Scientific
41764.HW 41764.HX 41764.GJ
41764.HWEA 303 EUR
41764.HW 41764.HX 41764.GJ
Molybdenum ≥99.95% (metals basis), foil, Thickness: 0.127 mm (0.005 in)
Molybdenum
Purity: 99.95% CAS No.: 7439-98-7 Physical Form: ≈19.5g/100x150mm, Temper: Hard MDL No.: MFCD00003465
Formula: Mo
MW: 95.94 g/mol
Boiling Pt: 3617 °C (1013 hPa)
Melting Pt: 2622 °C
Density: 10.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003465
CAS Number: 7439-98-7
EINECS: 231-107-2
Merck Index: 12,06317