Molybdenum ≥99.9% (metals basis), wire, annealed, Ø 1.58 mm (0.0625 in)
Supplier: Thermo Scientific
45055.G6 45055.G5
45055.G6EA 456 EUR
45055.G6 45055.G5
Molybdenum ≥99.9% (metals basis), wire, annealed, Ø 1.58 mm (0.0625 in)
Molybdenum
Purity: 99.9+% CAS No.: 7439-98-7 Physical Form: ≈20.2g/m MDL No.: MFCD00003465
Formula: Mo
MW: 95.94 g/mol
Boiling Pt: 3617 °C (1013 hPa)
Melting Pt: 2622 °C
Density: 10.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003465
CAS Number: 7439-98-7
EINECS: 231-107-2
Merck Index: 12,06317