Tungsten ≥99.97% (metals basis), slug, Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)
Supplier: Thermo Scientific
44252.22 44252.14
44252.22EA 182 EUR
44252.22 44252.14
Tungsten ≥99.97% (metals basis), slug, Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)
Tungsten
Purity: 99.97% CAS No.: 7440-33-7 Physical Form: ≈7.8g/slug MDL No.: MFCD00011461
Formula: W
MW: 183.84 g/mol
Boiling Pt: 5660 °C (1013 hPa)
Melting Pt: 3410 °C
Density: 19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9
Merck Index: 12,09945