Tungsten ≥99.9% (metals basis), powder -325 mesh
Supplier: Thermo Fisher Scientific
|
Warning
|
39749.22
39749.36
39749.A3
39749.22EA
72.6
EUR
39749.22
39749.36
39749.A3
Tungsten ≥99.9% (metals basis), powder -325 mesh
Tungsten
Purity: 99.9%
CAS No.: 7440-33-7
Appearance: Grey
UN No.: 3089
MDL No.: MFCD00011461
Formula:
W Storage Temperature: Ambient |
MDL Number:
MFCD00011461 CAS Number: 7440-33-7 EINECS: 231-143-9 UN: 3089 ADR: 4.1,III |