Manganese ≥99.6% (metals basis), powder APS <10 µm
Supplier: Thermo Fisher Scientific
Warning
45098.A1 45098.30 45098.18
45098.A1EA 269 EUR
45098.A1 45098.30 45098.18
Manganese ≥99.6% (metals basis), powder APS <10 µm
Manganese
Purity: 99.6% CAS No.: 7439-96-5 UN No.: 3089 MDL No.: MFCD00011111
Formula: Mn
MW: 54.94 g/mol
Density: 7.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011111
CAS Number: 7439-96-5
EINECS: 231-105-1
UN: 3089
ADR: 4.1 III