Manganese, powder, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ100
266132-1KG
266132-250G
SIAL266132-1KGEA
207
EUR
SIAL266132-1KG
SIAL266132-250G
Manganese, powder, Sigma-Aldrich®
Manganese
Formula:
Mn MW: 54.94 g/mol Density: 7.2 g/cm³ (20 °C) |
MDL Number:
MFCD00011111 CAS Number: 7439-96-5 EINECS: 231-105-1 UN: 3089 ADR: 4.1 III |