Manganese, powder, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ100

463728-100G 463728-25G
SIAL463728-100GEA 298 EUR
SIAL463728-100G SIAL463728-25G
Manganese, powder, Sigma-Aldrich®
Manganese
Formula: Mn
MW: 54.94 g/mol
Density: 7.2 g/cm³ (20 °C)
MDL Number: MFCD00011111
CAS Number: 7439-96-5
EINECS: 231-105-1
UN: 3089
ADR: 4.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR