Cobalt 99,5 Metals basis, powder 1.6 µm
Supplier: Thermo Fisher Scientific
Danger
10455.09 10455.22 10455.36
10455.09EA 95.1 EUR
10455.09 10455.22 10455.36
Cobalt 99,5 Metals basis, powder 1.6 µm
Cobalt
Purity: 99.8% CAS No.: 7440-48-4 Appearance: Black Odour: Odourless UN No.: 3089 MDL No.: MFCD00010935
Formula: Co
MW: 58.93 g/mol
Boiling Pt: 2870
Melting Pt: 1495
Storage Temperature: Ambient
MDL Number: MFCD00010935
CAS Number: 7440-48-4
EINECS: 231-158-0
UN: 3089
ADR: 4.1 II
Merck Index: 12,02488