Cobalt ≥99.995% (metals basis), wire, Puratronic®, Ø 0.5 mm (0.02 in)
Supplier: Thermo Fisher Scientific
Danger
10947.BW 10947.G2
10947.BWEA 195 EUR
10947.BW 10947.G2
Cobalt ≥99.995% (metals basis), wire, Puratronic®, Ø 0.5 mm (0.02 in)
Cobalt
Purity: 99.995% CAS No.: 7440-48-4 Physical Form: ≈1.75g/m Odour: Odourless MDL No.: MFCD00010935
Formula: Co
MW: 58.93 g/mol
Boiling Pt: 2870
Melting Pt: 1495
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010935
CAS Number: 7440-48-4
EINECS: 231-158-0
Merck Index: 12,02488