Cobalt 99,95 Metals basis, slug, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Supplier: Thermo Fisher Scientific
Danger
39671.14 39671.22
39671.14EA 148 EUR
39671.14 39671.22
Cobalt 99,95 Metals basis, slug, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Cobalt
Purity: 99.95% Physical Form: ≈1.8g/slug Odour: Odourless MDL No.: MFCD00010935
Formula: Co
MW: 58.93 g/mol
Boiling Pt: 2870
Melting Pt: 1495
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010935
CAS Number: 7440-48-4
EINECS: 231-158-0
Merck Index: 12,02488