Cobalt ≥99.9% (metals basis), foil 50x50 mm (2x2 in), Thickness: 0.127 mm (0.005 in)
Supplier: Thermo Fisher Scientific
Danger
39715.KG 39715.KJ
39715.KGEA 193 EUR
39715.KG 39715.KJ
Cobalt ≥99.9% (metals basis), foil 50x50 mm (2x2 in), Thickness: 0.127 mm (0.005 in)
Cobalt
Purity: 99.9% CAS No.: 7440-48-4 Physical Form: ≈2.83g/50x50mm Odour: Odourless MDL No.: MFCD00010935
Formula: Co
MW: 58.93 g/mol
Boiling Pt: 2870
Melting Pt: 1495
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010935
CAS Number: 7440-48-4
EINECS: 231-158-0
Merck Index: 12,02488