Cobalt 99,95 Metals basis, foil, Thickness: 0.1 mm (0.004 in)
Supplier: Thermo Fisher Scientific
Danger
42658.FF 42658.FI 42658.GH
42658.FFEA 83.9 EUR
42658.FF 42658.FI 42658.GH
Cobalt 99,95 Metals basis, foil, Thickness: 0.1 mm (0.004 in)
Cobalt
Purity: 99.95% CAS No.: 7440-48-4 Physical Form: ≈0.56g/25x25mm Odour: Odourless MDL No.: MFCD00010935
Formula: Co
MW: 58.93 g/mol
Boiling Pt: 2870
Melting Pt: 1495
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010935
CAS Number: 7440-48-4
EINECS: 231-158-0
Merck Index: 12,02488