Cobalt ≥99.95% (metals basis), foil, Thickness: 0.5 mm (0.02 in)
Supplier: Thermo Fisher Scientific
Danger
42659.FF 42659.FI 42659.GH
42659.FFEA 160 EUR
42659.FF 42659.FI 42659.GH
Cobalt ≥99.95% (metals basis), foil, Thickness: 0.5 mm (0.02 in)
Cobalt
Purity: 99.95% CAS No.: 7440-48-4 Physical Form: ≈2.8g/25x25mm Odour: Odourless MDL No.: MFCD00010935
Formula: Co
Boiling Pt: 2870 °C (1013 hPa)
Melting Pt: 1495 °C
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010935
CAS Number: 7440-48-4
EINECS: 231-158-0
Merck Index: 12,02488