Cobalt 99,95 Metals basis, foil, Thickness: 1.0 mm (0.04 in)
Supplier: Thermo Fisher Scientific
|
Danger
|
42660.FF
42660.FI
42660.FFEA
164
EUR
42660.FF
42660.FI
Cobalt 99,95 Metals basis, foil, Thickness: 1.0 mm (0.04 in)
Cobalt
Purity: 99.95%
CAS No.: 7440-48-4
Physical Form: ≈5.6g/25x25mm
Odour: Odourless
MDL No.: MFCD00010935
Formula:
Co MW: 58.93 g/mol Boiling Pt: 2870 Melting Pt: 1495 Density: 8.9 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010935 CAS Number: 7440-48-4 EINECS: 231-158-0 Merck Index: 12,02488 |