Cobalt, Sigma-Aldrich®
Supplier: Merck
Danger
MQ100
356867-2.2G 356867-8.8G
SIAL356867-2.2GEA 293 EUR
SIAL356867-2.2G SIAL356867-8.8G
Cobalt, Sigma-Aldrich®
Cobalt
Formula: Co
MW: 58.93 g/mol
Boiling Pt: 2870
Melting Pt: 1495
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010935
CAS Number: 7440-48-4
EINECS: 231-158-0
Merck Index: 12,02488