Iron 99,995 Metals basis, rods, Puratronic®, Ø 5 mm (0.2 in)
Supplier: Thermo Fisher Scientific
11442.DJ 11442.DO 11442.HT
11442.DJEA 257 EUR
11442.DJ 11442.DO 11442.HT
Iron 99,995 Metals basis, rods, Puratronic®, Ø 5 mm (0.2 in)
Iron
Purity: 99.995% CAS No.: 7439-89-6 Physical Form: ≈0.155g/mm MDL No.: MFCD00010999
Formula: Fe
MW: 55.85 g/mol
Boiling Pt: 3000
Melting Pt: 1535
Density: 7.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010999
CAS Number: 7439-89-6
EINECS: 231-096-4
Merck Index: 13,05109