Iron 99,99 Metals basis, wire, Ø 1.0 mm (0.04 in)
Supplier: Thermo Fisher Scientific
40497.BW 40497.CB
40497.BWEA 156 EUR
40497.BW 40497.CB
Iron 99,99 Metals basis, wire, Ø 1.0 mm (0.04 in)
Iron
Purity: 99.99% CAS No.: 7439-89-6 Physical Form: ≈6.2g/m MDL No.: MFCD00010999
Formula: Fe
MW: 55.85 g/mol
Boiling Pt: 3000
Melting Pt: 1535
Density: 7.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010999
CAS Number: 7439-89-6
EINECS: 231-096-4
Merck Index: 13,05109