Iron, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
267945-1KG
267945-250G
SIAL267945-1KGEA
145
EUR
SIAL267945-1KG
SIAL267945-250G
Iron, Sigma-Aldrich®
Iron
Formula:
Fe Boiling Pt: 3000 °C (1013 hPa) Melting Pt: 1535 °C Density: 7.86 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010999 CAS Number: 7439-89-6 EINECS: 231-096-4 Merck Index: 13,05109 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...