Iron, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

413054-25G 413054-5G
SIAL413054-25GEA 302 EUR
SIAL413054-25G SIAL413054-5G
Iron, Sigma-Aldrich®
Iron
Formula: Fe
Boiling Pt: 3000 °C (1013 hPa)
Melting Pt: 1535 °C
Density: 7.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010999
CAS Number: 7439-89-6
EINECS: 231-096-4
Merck Index: 13,05109

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR