Iron, powder, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ100
209309-500G
209309-2KG
SIAL209309-500GEA
81
EUR
SIAL209309-500G
SIAL209309-2KG
Iron, powder, Sigma-Aldrich®
Iron
Formula:
Fe Boiling Pt: 3000 °C (1013 hPa) Melting Pt: 1536 °C Density: 7.86 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010999 CAS Number: 7439-89-6 EINECS: 231-096-4 UN: 3089 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...