Iridium ≥99.8% (metals basis), wire, Ø 1.0 mm (0.04 in)
Supplier: Thermo Fisher Scientific
11429.BO 11429.BQ
11429.BOEA 379 EUR
11429.BO 11429.BQ
Iridium ≥99.8% (metals basis), wire, Ø 1.0 mm (0.04 in)
Iridium
Purity: 99.8% CAS No.: 7439-88-5 Physical Form: ≈17.8g/m MDL No.: MFCD00011062
Formula: Ir
Boiling Pt: 5300 °C (1013 hPa)
Melting Pt: 2450 °C
Storage Temperature: Ambient
MDL Number: MFCD00011062
CAS Number: 7439-88-5
EINECS: 231-095-9