Iridium ≥99.95% (metals basis), sponge -100 mesh
Supplier: Thermo Fisher Scientific
Warning
12069.02 12069.04 12069.09
12069.02EA 0 EUR
12069.02 12069.04 12069.09
Iridium ≥99.95% (metals basis), sponge -100 mesh
Iridium
Purity: 99.95% CAS No.: 7439-88-5 UN No.: 3089 MDL No.: MFCD00011062
Formula: Ir
MW: 192.22 g/mol
Boiling Pt: 5300
Melting Pt: 2450
Storage Temperature: Ambient
MDL Number: MFCD00011062
CAS Number: 7439-88-5
EINECS: 231-095-9
UN: 3089
ADR: 4.1 III