Iridium ≥99.9% (metals basis), powder -325 mesh
Supplier: Thermo Scientific
Warning
39684.02 39684.04
39684.02EA 457 EUR
39684.02 39684.04
Iridium ≥99.9% (metals basis), powder -325 mesh
Iridium
Purity: 99.9% CAS No.: 7439-88-5 Molecular Formula: Ir Physical Form: Powder/Sponge UN No.: 3089 MDL No.: MFCD00011062
Formula: Ir
MW: 192.22 g/mol
Boiling Pt: 4500 °C (1013 hPa)
Melting Pt: 2443 °C
Density: 22.65 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011062
CAS Number: 7439-88-5
EINECS: 231-095-9
UN: 3089
ADR: 4.1,III
Merck Index: 12,05098