Nickel 99 Metals basis, foil, annealed, Thickness: 0.127 mm
Supplier: Thermo Fisher Scientific
Danger
10595.CH 10595.CJ 10595.RX
10595.CHEA 136 EUR
10595.CH 10595.CJ 10595.RX
Nickel 99 Metals basis, foil, annealed, Thickness: 0.127 mm
Nickel
Purity: 99+% CAS No.: 7440-02-0 Physical Form: ≈67.8g/20x30cm, Temper: soft Odour: Odourless MDL No.: MFCD00011137
Formula: Ni
MW: 58.69 g/mol
Boiling Pt: 3007
Melting Pt: 1453
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1 III
Merck Index: 12,06582