Nickel 99,994 Metals basis, wire, Puratronic®, Ø 0.25 mm (0.01 in)
Supplier: Thermo Fisher Scientific
Danger
10931.G2 10931.G6 10931.H2
10931.G2EA 148 EUR
10931.G2 10931.G6 10931.H2
Nickel 99,994 Metals basis, wire, Puratronic®, Ø 0.25 mm (0.01 in)
Nickel
Purity: 99.994% CAS No.: 7440-02-0 Physical Form: ≈0.44g/m Odour: Odourless MDL No.: MFCD00011137
Formula: Ni
MW: 58.69 g/mol
Boiling Pt: 3007
Melting Pt: 1453
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1 III
Merck Index: 12,06582