Nickel ≥99.5% (metals basis), foil, annealed, Thickness: 0.787 mm (0.031 in)
Supplier: Thermo Fisher Scientific
Danger
14193.GH 14193.HP
14193.GHEA 68 EUR
14193.GH 14193.HP
Nickel ≥99.5% (metals basis), foil, annealed, Thickness: 0.787 mm (0.031 in)
Nickel
Purity: 99.5% CAS No.: 7440-02-0 Physical Form: ≈70g/100x100mm Odour: Odourless MDL No.: MFCD00011137
Formula: Ni
Boiling Pt: 3007 °C (1013 hPa)
Melting Pt: 1453 °C
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1,III
Merck Index: 12,06582